missing translation for 'onlineSavingsMsg'
Learn More

Androst-4-ene-3,6,17-trione, TRC

Product Code. 18083110
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18083110 50 mg 50mg
18064010 5 mg 5mg
18023320 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18083110 Supplier Toronto Research Chemicals Supplier No. TRCA63756550MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2243-06-3
Molecular Formula C19 H24 O3
Molecular Weight (g/mol) 300.39
Synonym Androst-4-ene-3,6,17-trione, 4-Androstene-3,6,17-trione
IUPAC Name (8R,9S,10R,13S,14S)-10,13-dimethyl-1,2,7,8,9,11,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,6,17-trione
SMILES C[C@]12CC[C@H]3[C@@H](CC(=O)C4=CC(=O)CC[C@]34C)[C@@H]1CCC2=O

Specifications

Analyte or Component Names Androst-4-ene-3,6,17-trione
Shipping Condition Room Temperature
InChI Formula InChI=1S/C19H24O3/c1-18-7-5-11(20)9-15(18)16(21)10-12-13-3-4-17(22)19(13,2)8-6-14(12)18/h9,12-14H,3-8,10H2,1-2H3/t12-,13-,14-,18+,19-/m0/s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 50 mg
Formula Weight 300.1725
Percent Purity >95%
Chemical Name or Material Androst-4-ene-3,6,17-trione
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.