missing translation for 'onlineSavingsMsg'
Learn More

Berthold Technologies Anesthesia Unit for NightOWL LB 983

Product Code. 31908717 Shop All Berthold Technologies Products
Change view
Click to view available options
Color:
Black
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
31908717 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31908717 Supplier Berthold Technologies Supplier No. S.A.50/24 230/50
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Required to keep mice immobile and allow imaging of living animals in the NightOWL II.

  • Complete with nozzle part and tray for up to 5 mice.
  • Additional trays can be purchased separately.

Specifications

For Use With (Application) Animal Inhalant Anesthesia
For Use With (Equipment) NightOWL II
Voltage 230 V
Certifications CE
Animal Mouse
Type Anesthesia Machine
Color Black
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.