missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiomotin like 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
285.00 € - 500.00 €
Specifications
| Antigen | Angiomotin like 1 |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18489381
|
Novus Biologicals
NBP1-91665-25ul |
25 μL |
285.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18084947
|
Novus Biologicals
NBP1-91665 |
0.1 mL |
500.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Angiomotin like 1 Polyclonal specifically detects Angiomotin like 1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Angiomotin like 1 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q8IY63 | |
| 154810 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SPSACYSPSSPVQVLEDSTYFSPDFQLYSGRHETSALTVEATSSIREKVVEDPLCNFHSPNFLRISEVEMRGSEDAAAGTVLQRLIQEQLRYGTPTENMNLLAIQHQATGSAGPAHPTNNFSSTENLTQEDPQMVYQSARQEPQGQEHQV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| angiomotin like 1, angiomotin-like protein 1, JEAP, junction-enriched and associated protein | |
| AMOTL1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title