Learn More
Angiopoietin-1 Antibody (S06-1F9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19492-100ul
 View more versions of this product
Product Code. 18673695
  • 645.00 € / 100µL
Description
Angiopoietin-1 Monoclonal antibody specifically detects Angiopoietin-1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, RNA Inhibition
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Angiopoietin-1
Monoclonal
0.3 mg/mL
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:1000, RNA Inhibition
AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, KIAA0003
A synthetic peptide of human Angiopoietin-1 (Uniprot # Q15389)
100 μg
Angiogenesis, Cancer, Cell Cycle and Replication
284
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, ELISA, Immunofluorescence, Immunoprecipitation, Inhibition Assays
S06-1F9
Unconjugated
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Rat
Purified