missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Angiopoietin 3 Monoclonal Antibody (113504)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561810
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Angiopoietin-1 (Ang-1) is a secreted ligand for Tie-2, a cell surface receptor tyrosine kinase expressed in endothelial and hemopoietic cells. Ang-1 is an angiogenic factor that mediates blood vessel maturation and may be involved in endothelial development. A related protein, angiopoietin-2 (Ang-2), is a naturally occurring antagonist of Ang-1 activation of Tie-2. In adult tissue, Ang-2 expression is restricted to sites of vascular remodeling. Ang-3 and Ang-4 represent the mouse and human counterparts of the same gene locus. The structural divergence of Ang-3 and Ang-4 cause their divergent functions. Ang-3 and Ang-4 have very different distributions in their respective species, and Ang-3 appears to act as an antagonist while Ang-4 appears to function as an agonist. Ang-3 and Ang-4 share all the main structural characteristics of Ang-1 and Ang-2 and are homologous throughout the signal peptide, N-terminal region, coiled-coil segment and Fibrinogen-like domain.
Specifications
| Angiopoietin 3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9WVH6 | |
| ANGPT4 | |
| E. coli-derived recombinant mouse Angiopoietin-3. | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 113504 | |
| Unconjugated | |
| ANGPT4 | |
| AGP4; Agpt4; ANG3; ANG-3; ANG4; ANG-4; angiopoietin 3; angiopoietin 4; Angiopoietin3; angiopoietin-3; Angiopoietin4; Angiopoietin-4; Angpt4 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 11602 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction