missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ANGPTL1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 30533626
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30533626 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30533626 Supplier Invitrogen™ Supplier No. PA5148573

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

Angiopoietins are members of the vascular endothelial growth factor family and the only known growth factors largely specific for vascular endothelium. Angiopoietin-1, angiopoietin-2, and angiopoietin-4 participate in the formation of blood vessels. The protein encoded by this gene is another member of the angiopoietin family that is widely expressed in adult tissues with mRNA levels highest in highly vascularized tissues. This protein was found to be a secretory protein that does not act as an endothelial cell mitogen in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANGPTL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene Angptl1
Gene Accession No. O95841
Gene Alias 2810039D03Rik; ANG3; ANG-3; angioarrestin; angiopoietin 3; angiopoietin like 1; angiopoietin Y1; angiopoietin-3; angiopoietin-like 1; angiopoietin-like protein 1; Angiopoietin-related protein 1; ANGPT3; ANGPTL1; AngY; ARP1; dJ595C2.2; dJ595C2.2 (angiopoietin Y1); PSEC0154; UNQ162; UNQ162/PRO188
Gene Symbols Angptl1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-like 1/ANGPTL1, Ala39-Asp491.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9068
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.