missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL7/MCP-3/MARC ELISA Kit (Colorimetric)

Product Code. 18608870 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18608870 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18608870 Supplier R&D Systems™ Supplier No. KA2199
  • ONLINE EXCLUSIVE PRICE
    1340.27 €
    1131.90 € / 1 sets
    Save 208.37 €
    15% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems.

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 8 to 2000 pg/mL
Product Line Novus
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Lysates, Plasma, Serum
For Use With (Application) ELISA, Quantification
Gene ID (Entrez) 6354
Gene Symbol CCL7
Quantity 1 Kit
Research Discipline Cancer
Storage Requirements Store at 4°C.
Synonym C-C motif chemokine 7, chemokine (C-C motif) ligand 7, FIC, MARC, MCP3Monocyte chemotactic protein 3, MCP-3 Small-inducible cytokine A7, MGC138465, Monocyte chemoattractant protein 3, NC28SCYA6, SCYA7MGC138463, small inducible cytokine A7 (monocyte chemotactic protein 3)
Target Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.