Learn More
Anillin Antibody (CL0303) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52908AF647
Product Code. 30302019
  • 770.00 € / 0.10mL
Description
Anillin Monoclonal antibody specifically detects Anillin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Anillin
Monoclonal
Alexa Fluor 647
actin-binding protein anillin, ANILLIN, anillin (Drosophila Scraps homolog), actin binding protein, anillin, actin binding protein, anillin, actin binding protein (scraps homolog, Drosophila), DKFZp779A055, Drosophila Scraps homolog, scra, Scraps
This antibody was developed using a recombinant protein derived from Q9NQW6, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence
CL0303
50mM Sodium Borate
Mouse
Protein A purified
RUO
54443
Store at 4⪚C in the dark.
IgG1