missing translation for 'onlineSavingsMsg'
Learn More

Anilofos 1000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30309877
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309877 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309877 Supplier Dr. Ehrenstorfer Supplier No. DREA10264000ME1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Anilofos
CAS 64249-01-0
Analyte or Component Names Anilofos
Shipping Condition +20°C
Molecular Formula C13 H19 Cl N O3 P S2
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C13H19ClNO3PS2/c1-10(2)15(12-7-5-11(14)6-8-12)13(16)9-21-19(20,17-3)18-4/h5-8,10H,9H2,1-4H3
SMILES COP(=S)(OC)SCC(=O)N(C(C)C)c1ccc(Cl)cc1
IUPAC Name N-(4-chlorophenyl)-2-dimethoxyphosphinothioylsulfanyl-N-propan-2-ylacetamide
Solvent or Matrix Methanol
Molecular Weight (g/mol) 367.85
Quantity 1 mL
Formula Weight 367.0232 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.