missing translation for 'onlineSavingsMsg'
Learn More

Anisaldehyde, TRC

Product Code. 30136857
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136857 100 g 100g
30133471 250 g 250g
30007649 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136857 Supplier Toronto Research Chemicals Supplier No. TRCA673200100G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-11-5
Molecular Formula C8 H8 O2
Molecular Weight (g/mol) 136.15
Synonym Benzaldehyde, 4-methoxy- (9CI, ACI), 4-Methoxybenzaldehyde (ACI), p-Anisaldehyde (8CI), 4-Anisaldehyde, Anisaldehyde, Anisic aldehyde, Aubepine, Crategine, NSC 5590, Obepin, p-Anisic aldehyde, p-Formylanisole, p-Methoxybenzaldehyde
IUPAC Name 4-methoxybenzaldehyde
SMILES COc1ccc(C=O)cc1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C8H8O2/c1-10-8-4-2-7(6-9)3-5-8/h2-6H,1H3
Formula Weight 136.0524
Chemical Name or Material 4-Anisaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.