missing translation for 'onlineSavingsMsg'
Learn More

Anisoin, 95%

Product Code. 10773631 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10773631 100 g Plastic bottle 100g
10657484 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10773631 Supplier Thermo Scientific Acros Supplier No. 104841000
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 119-52-8 | C16H16O4 | 272.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-52-8
Molecular Formula C16H16O4
Molecular Weight (g/mol) 272.30
MDL Number MFCD00008411
InChI Key LRRQSCPPOIUNGX-UHFFFAOYNA-N
Synonym anisoin, 2-hydroxy-1,2-bis 4-methoxyphenyl ethanone, 4,4'-dimethoxybenzoin, p-anisoin, ethanone, 2-hydroxy-1,2-bis 4-methoxyphenyl, benzoin, 4,4'-dimethoxy, p,p'-dimethoxybenzoin, benzoin,4'-dimethoxy, 4,4'-anisoin, 1,2-bis 4-methoxyphenyl-2-hydroxyethan-1-one
PubChem CID 95415
IUPAC Name 2-hydroxy-1,2-bis(4-methoxyphenyl)ethanone
SMILES COC1=CC=C(C=C1)C(O)C(=O)C1=CC=C(OC)C=C1

Specifications

Melting Point 109°C to 114°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 08, 423
Formula Weight 272.29
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Anisoin, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.