missing translation for 'onlineSavingsMsg'
Learn More

Anisole, ReagentPlus™, 99%, Solstice

Product Code. p-7120201
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
1 L
2.5 L
20 L
250mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15691120 250 mL Glass Bottle 250mL
15661120 1 L Glass Bottle 1 L
15671120 2.5 L Glass Bottle 2.5 L
15681120 20 L Stainless Steel Can 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15691120 Supplier Honeywell Research Chemicals Supplier No. 123226250ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-66-3
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00008354
InChI Key RDOXTESZEPMUJZ-UHFFFAOYSA-N
Synonym methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene
PubChem CID 7519
ChEBI CHEBI:16579
IUPAC Name anisole
SMILES COC1=CC=CC=C1

Specifications

Melting Point -37°C
Color Colorless
Density 0.995g/mL at 25°C
Boiling Point 154°C
Flash Point 51°C (123.8°F)
Packaging Glass Bottle
Linear Formula CH3OC6H5
Refractive Index n20/D 1.516(lit.)
Quantity 250 mL
UN Number UN2222
Beilstein 506892
Vapor Density 3.7 (vs air)
Formula Weight 108.14g/mol
Percent Purity 99%
Grade ReagentPlus
Physical Form Liquid
Chemical Name or Material Anisole
Show More Show Less
compliance-icons
Product Identifier
  • Anisole
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.