missing translation for 'onlineSavingsMsg'
Learn More

Anisomycin, MedChemExpress

Product Code. 16427678
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16427678

Brand: Medchem Express HY18982

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Anisomycin is a potent protein synthesis inhibitor which interferes with protein and DNA synthesis by inhibiting peptidyl transferase or the 80S ribosome system. Anisomycin is a JNK activator, which increases phospho-JNK. Anisomycin is a bacterial antibiotic.

Anisomycin is a potent protein synthesis inhibitor which interferes with protein and DNA synthesis by inhibiting peptidyl transferase or the 80S ribosome system. Anisomycin is a JNK activator, which increases phospho-JNK. Anisomycin is a bacterial antibiotic.

Specifications

Chemical Name or Material Anisomycin
Physical Form Solid
CAS 22862-76-6
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C14H19NO4
For Use With (Application) Cancer-Kinase/protease
Synonym Flagecidin Wuningmeisu C
Solubility Information DMSO : ≥ 50 mg/mL (188.46 mM)
SMILES O[C@@H]1[C@@H](OC(C)=O)[C@@H](CC2=CC=C(OC)C=C2)NC1
Molecular Weight (g/mol) 265.31
Formula Weight 265.31
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.