missing translation for 'onlineSavingsMsg'
Learn More

Anisonitrile, 99%

Product Code. 10021850 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10021850 25 g Glass bottle 25g
10133160 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10021850 Supplier Thermo Scientific Acros Supplier No. 158050250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 874-90-8 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874-90-8
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00001818
InChI Key XDJAAZYHCCRJOK-UHFFFAOYSA-N
Synonym anisonitrile, p-anisonitrile, 4-cyanoanisole, benzonitrile, 4-methoxy, p-methoxybenzonitrile, 4-anisonitrile, p-anisylnitrile, p-cyanoanisole, p-methoxyphenyl cyanide, 4-methoxy benzonitrile
PubChem CID 70129
IUPAC Name 4-methoxybenzonitrile
SMILES COC1=CC=C(C=C1)C#N

Specifications

Melting Point 57°C to 60°C
Color White to Beige
Boiling Point 256°C to 257°C (765.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3OC6H4CN
Quantity 25 g
Beilstein 10, 168
Solubility Information Solubility in water: soluble
Formula Weight 133.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Anisonitrile, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Anisonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.