missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKHD1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92751-0.02ml
This item is not returnable.
View return policy
Description
ANKHD1 Polyclonal antibody specifically detects ANKHD1 in Human samples. It is validated for Western Blot
Specifications
| ANKHD1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ankyrin repeat and KH domain containing 1, FLJ10042, FLJ11979, FLJ14127, hMASK, KIAA1085single KH-domain homolog, MASKFLJ20288, Multiple ankyrin repeats single KH domain | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1620-1800 of human ANKHD1 (NP_060217.1). VNSSKYPSLLLHSQEEKTSTATSKTQTRLEGEVTPNSLSTSYKTVSLPLSSPNIKLNLTSPKRGQKREEGWKEVVRRSKKLSVPASVVSRIMGRGGCNITAIQDVTGAHIDVDKQKDKNGERMITIRGGTESTRYAVQLINALIQDPAKELEDLIPKNHIRTPASTKSIHANFSSGVGTTA | |
| 0.02 mL | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Immunology | |
| 54882 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction