missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium-sensing R/CaSR Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41271B
This item is not returnable.
View return policy
Description
Calcium-sensing R/CaSR Polyclonal antibody specifically detects Calcium-sensing R/CaSR in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Calcium-sensing R/CaSR | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Biotin | |
| calcium-sensing receptor, CAR, CaSR, EIG8, extracellular calcium-sensing receptor, FHH, FIH, GPRC2AHHC, HHC1, hypocalciuric hypercalcemia 1, NSHPT, parathyroid Ca(2+)-sensing receptor 1, Parathyroid cell calcium-sensing receptor, PCaR1, PCAR1MGC138441 | |
| Antibody was raised against a 15 amino acid peptide near the amino terminus of human CASR. The immunogen is located within the first 50 amino acids of CASR. Amino Acid Squence: TSAYGPDQRAQKKGD | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Extracellular Matrix, GPCR, Growth and Development, Lipid and Metabolism, Neuroscience, Neurotransmission, Signal Transduction | |
| 846 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction