missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKLE2 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-41145AF647
This item is not returnable.
View return policy
Description
ANKLE2 Polyclonal antibody specifically detects ANKLE2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| ANKLE2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| Alexa Fluor 647 | |
| ankyrin repeat and LEM domain containing 2, ankyrin repeat and LEM domain-containing protein 2, FLJ22280, FLJ36132, KIAA0692, LEM domain containing 7, LEMD7 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the center of human ANKLE2. The immunogen is located within amino acids 280 - 330 of ANKLE2. Amino Acid Squence: CSESETVNKERANSYKNPRT | |
| 0.1 mL | |
| Cell Biology | |
| 23141 | |
| Store at 4°C in the dark. | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction