missing translation for 'onlineSavingsMsg'
Learn More

ANKRD1 Mouse, anti-Human, CoraLite Plus 555, Clone: 1F11F1, Proteintech

Product Code. 18005145 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18005145 100 μL 100µL
18079795 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18005145 Supplier Proteintech Supplier No. CL55567775100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The ANKRD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ANKRD1. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The ANKRD1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 555.

Specifications

Antigen ANKRD1
Applications Immunofluorescence
Clone 1F11F1
Concentration 1000 μg/mL
Conjugate CoraLite Plus 555
Gene Symbols ALRP, ANKRD1, bA320F15.2, C 193, C193, Cardiac ankyrin repeat protein, CARP, CVARP, HA1A2, MCARP
Host Species Mouse
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 27063
Target Species Human, Mouse, Rat, Rabbit, Pig
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Antibody Molecular Weight 155 kDa
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.