missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD15 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92635-0.02ml
This item is not returnable.
View return policy
Description
ANKRD15 Polyclonal antibody specifically detects ANKRD15 in Mouse samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| ANKRD15 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| ANKRD15ankyrin repeat domain 15, Ankyrin repeat domain-containing protein 15, KANKDKFZp451G231, KIAA0172MGC43128, Kidney ankyrin repeat-containing protein, KN motif and ankyrin repeat domain-containing protein 1, KN motif and ankyrin repeat domains 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 900-1000 of human KANK1 (NP_055973.2). GNYLGYTCKCGGLQSGSPLSSQTSQPEQEVGTSEGKPISSLDAFPTQEGTLSPVNLTDDQIAAGLYACTNNESTLKSIMKKKDGNKDSNGAKKNLQFVGIN | |
| 0.02 mL | |
| Cell Cycle and Replication | |
| 23189 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction