missing translation for 'onlineSavingsMsg'
Learn More

ANKRD15 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

198.00 € - 462.00 €

Specifications

Antigen ANKRD15
Dilution Immunocytochemistry/ Immunofluorescence 1:50-1:200
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18673782
View Documents
Novus Biologicals
NBP2-92635-0.02ml
0.02 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
18687352
View Documents
Novus Biologicals
NBP2-92635-0.1ml
0.1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

ANKRD15 Polyclonal antibody specifically detects ANKRD15 in Mouse samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

ANKRD15
Immunofluorescence
Unconjugated
Rabbit
Cell Cycle and Replication
PBS with 50% glycerol, pH7.3.
23189
IgG
Affinity purified
Immunocytochemistry/ Immunofluorescence 1:50-1:200
Polyclonal
Purified
RUO
Mouse
ANKRD15ankyrin repeat domain 15, Ankyrin repeat domain-containing protein 15, KANKDKFZp451G231, KIAA0172MGC43128, Kidney ankyrin repeat-containing protein, KN motif and ankyrin repeat domain-containing protein 1, KN motif and ankyrin repeat domains 1
A synthetic peptide corresponding to a sequence within amino acids 900-1000 of human KANK1 (NP_055973.2). GNYLGYTCKCGGLQSGSPLSSQTSQPEQEVGTSEGKPISSLDAFPTQEGTLSPVNLTDDQIAAGLYACTNNESTLKSIMKKKDGNKDSNGAKKNLQFVGIN
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.