missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ANKRD36C Recombinant Protein Antigen
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Beskrivelse
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human ANKRD36C. Source: E.coli Amino Acid Sequence: KDEQISGTVSSQKQPALKATSDKKDSVSNIPTEIKDGQQSGTVSSQKQLAWKATSVKKDSVSNIATEIKDGQIRGTVS The ANKRD36C Recombinant Protein Antigen is derived from E. coli. The ANKRD36C Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Tekniske data
Tekniske data
| Gene ID (Entrez) | 400986 |
| Purification Method | >80% by SDS-PAGE and Coomassie blue staining |
| Common Name | ANKRD36C Recombinant Protein Antigen |
| Content And Storage | −20°C, avoid freeze-thaw cycles |
| Formulation | PBS and 1M Urea, pH 7.4 |
| For Use With (Application) | Blocking/Neutralizing, Control |
| Gene Alias | Ankyrin Repeat Domain 36C, Protein Immuno-Reactive With Anti-PTH Polyclonal Antibodies |
| Gene Symbol | N/A |
| Label Type | Unlabeled |
| Product Type | Recombinant Protein Antigen |
| Vis mere |
For research use only.
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion