missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD54 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49652-25ul
This item is not returnable.
View return policy
Description
ANKRD54 Polyclonal antibody specifically detects ANKRD54 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ANKRD54 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| ankyrin repeat domain 54, LIARankyrin repeat domain-containing protein 54 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RSGHSSSEGECAVAPEPLTDAEGLFSFADFGSALGGGGAGLSGRASGGAQSPLRYLHVLWQQDAEP | |
| 25 μL | |
| Signal Transduction | |
| 129138 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction