missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ankyrin 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
321.00 € - 536.00 €
Specifications
| Antigen | Ankyrin 3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500, Immunocytochemistry/ Immunofluorescence 1:1000 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18646956
|
Novus Biologicals
NBP3-05550-100ul |
100 μg |
536.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18649346
|
Novus Biologicals
NBP3-05550-50ul |
50 μg |
321.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ankyrin 3 Polyclonal antibody specifically detects Ankyrin 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Ankyrin 3 | |
| Western Blot 1:500, Immunocytochemistry/ Immunofluorescence 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ankyrin G, Ankyrin G119, ankyrin-3, ankyrin-G, Node Of Ranvier | |
| The C-terminal 398 amino acids of human Ankyrin 3 expressed in and purified from E. coli. | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cellular Signaling, Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers | |
| 50% PBS, 50% glycerol | |
| 288 | |
| IgG1 | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title