missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A1 Antibody (CL0199) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52875V
This item is not returnable.
View return policy
Description
Annexin A1 Monoclonal antibody specifically detects Annexin A1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| Annexin A1 | |
| Monoclonal | |
| DyLight 405 | |
| annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein | |
| This antibody was developed using a recombinant protein derived from P04083, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Protein Kinase, Signal Transduction | |
| 301 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL0199 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction