missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 18 publications
Brand: Novus Biologicals NBP1-90156-25ul
This item is not returnable.
View return policy
Description
Annexin A10 Polyclonal specifically detects Annexin A10 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Annexin A10 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Simple Western 1:20, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:200-1:500 | |
| Q9UJ72 | |
| ANXA10 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PPLYDAHELWHAMKGVGTDENCLIEILASRTNGEIFQMREAYCLQYSNNLQEDIYSETSGHFRDTLMNLVQGTREEGYTDPAMAAQDAMVLWEACQQKTGEHKTMLQMILCNK | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| annexin 14, annexin A10, annexin-10, annexin-14, ANX14Annexin-10 | |
| Rabbit | |
| 37 kDa | |
| 25 μL | |
| Signal Transduction | |
| 11199 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction