missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Annexin A2 Recombinant Rabbit Monoclonal Antibody (10H9)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550047
This item is not returnable.
View return policy
Description
Annexin A2 Recombinant Monoclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions as an autocrine factor which heightens osteoclast formation and bone resorption. This gene has three pseudogenes located on chromosomes 4, 9 and 10, respectively. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| Annexin A2 | |
| Recombinant Monoclonal | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P07355 | |
| ANXA2 | |
| A synthesized peptide derived from human ANXA2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 10H9 | |
| Unconjugated | |
| ANXA2 | |
| 33-kDa calcimedin; 33-kDa lymphocyte Ca>2+>- binding protein; 33kDa lymphocyte Ca>2+>- binding protein; 36-kDa calelectrin; annexin 2; annexin A2; annexin A2 isoform 2; annexin II; Annexin-2; ANX2; ANX2L4; ANXA2; AW215814; CAL1H; calpactin I heavy chain; calpactin I heavy chain (p36); calpactin I heavy polypeptide; calpactin-1 heavy chain; chromobindin 8; chromobindin-8; epididymis secretory protein Li 270; HEL-S-270; LIP2; lipocortin II; LPC2; LPC2D; p36; PAP-IV; Placental anticoagulant protein IV; protein I | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 302 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction