Learn More
Invitrogen™ Annexin A4 Recombinant Rabbit Monoclonal Antibody (24GB4715)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557333
 View more versions of this product
Product Code. 30275938
  • 564.00 € / 100µL
Description
Annexin A4 Recombinant Monoclonal Antibody for Western Blot, Flow
Annexin IV (ANX4) belongs to the annexin family of calcium-dependent phospholipid binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. ANX4 has 45 to 59% identity with other members of its family and shares a similar size and exon-intron organization. Isolated from human placenta, ANX4 encodes a protein that has possible interactions with ATP, and has in vitro anticoagulant activity and also inhibits phospholipase A2 activity. ANX4 is almost exclusively expressed in epithelial cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Annexin A4
Recombinant Monoclonal
Unconjugated
ANXA4
35-beta calcimedin; 36 kDa zymogen granule membrane-associated protein; AI265406; AIV; annexin 4; annexin A4; Annexin IV; annexin IV (placental anticoagulant protein II); annexin-4; ANX IV; ANX4; ANXA 4; Anxa4; ANXIV; AW106930; Carbohydrate-binding protein p33/p41; chromobindin-4; endonexin; endonexin I; epididymis secretory protein Li 274; HEL-S-274; Lipocortin IV; P32.5; p33/41 (annexin IV); PAP-II; PIG28; Placental anticoagulant protein II; PP4-X; proliferation-inducing gene 28; proliferation-inducing protein 28; Protein II; unnamed protein product; Xanx-4; ZAP 36/annexin IV; ZAP36; zymogen granule membrane associated protein
Rabbit
Affinity chromatography
RUO
11746, 307, 79124
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4715
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P09525, P55260, P97429
ANXA4
Recombinant protein of human Annexin-4/ANXA4 (aa 2-319).
100 μL
Primary
Human, Mouse, Rat