Learn More
Invitrogen™ Annexin A7 Recombinant Rabbit Monoclonal Antibody (23GB1235), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA554183
 View more versions of this product
Product Code. 30088277
  • 207.00 € / 20µL
Description
Calcium/phospholipid-binding protein which promotes membrane fusion and is involved in exocytosis. Annexin VII is a member of the annexin family of calcium-dependent phospholipid binding proteins. The Annexin VII gene contains 14 exons and spans approxima.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Annexin A7
Recombinant Monoclonal
Unconjugated
ANXA7
AI265384; AI316497; Annexin A7; annexin VII; Annexin-7; Anx7; ANXA7; Interleukin-8; OK/SW-cl.95; RP11-537A6.8; SNX; SYNEXIN
Rabbit
Affinity chromatography
RUO
11750, 155423, 310
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB1235
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P20073, Q07076
ANXA7
A synthesized peptide derived from human Annexin A7 (1-150AA).
20 μL
Primary
Human, Mouse, Rat