missing translation for 'onlineSavingsMsg'
Learn More

ANO2 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

204.00 € - 474.00 €

Specifications

Antigen ANO2
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin
Applications Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18610792
View Documents
Novus Biologicals
NBP2-92888-0.02ml
0.02 mL
204.00 €
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18689691
View Documents
Novus Biologicals
NBP2-92888-0.1ml
0.1 mL
474.00 €
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ANO2 Polyclonal antibody specifically detects ANO2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

ANO2
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Neuroscience
PBS with 50% glycerol, pH7.3.
57101
IgG
Affinity purified
Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin
Polyclonal
Purified
RUO
Human, Mouse, Rat
anoctamin 2, anoctamin-2, DKFZp434P102, TMEM16B, transmembrane protein 16B (eight membrane-spanning domains)
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human ANO2 (NP_001265526.1). MPEDIPLLPGSPRRLSPQAGSRGGQGPKHGQQCLKMPGPRAPGLQGGSNRDPGQPCGGESTRSSSVINNYLDANEPVSLEARLSRMHFHDSQRKVDYVLAYHYRKRGVHLAQGFPGHSLAIVSNGETGKEPHAGGPGDIELGPLDALEEERKEQREEFEHNLMEAGLELEKDLENKSQGSIFVRIHAPWQVLAREAEFLK
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.