Learn More
ANO2 Antibody (CL10632) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-07992V
Product Code. 30300773
  • 737.00 € / 0.10mL
Description
ANO2 Monoclonal antibody specifically detects ANO2 in Human,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ANO2
Monoclonal
DyLight 405
anoctamin 2, anoctamin-2, DKFZp434P102, TMEM16B, transmembrane protein 16B (eight membrane-spanning domains)
This antibody was developed using a recombinant protein derived from Q9NQ90, with the exact immunogen sequence remaining proprietary.
0.1 mL
Neuroscience
57101
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL10632
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Rat
Purified