missing translation for 'onlineSavingsMsg'
Learn More

Antazoline Hydrochloride, TRC

Product Code. 30135664
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Unit Size:
100g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135664 1 g 1g
30133367 100 g 100g
17954465 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135664 Supplier Toronto Research Chemicals Supplier No. TRCA6784051G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2508-72-7
Molecular Formula C17 H19 N3 . Cl H
Molecular Weight (g/mol) 301.81
Synonym 1H-Imidazole-2-methanamine, 4,5-dihydro-N-phenyl-N-(phenylmethyl)-, hydrochloride (1:1), 1H-Imidazole-2-methanamine, 4,5-dihydro-N-phenyl-N-(phenylmethyl)-, monohydrochloride (9CI), 2-Imidazoline, 2-[(N-benzylanilino)methyl]-, monohydrochloride (8CI), 2-(N-Benzylanilinomethyl)-2-imidazoline hydrochloride, 5512M, Antastan, Antasten, Antazoline chloride, Antazoline hydrochloride, Antihistal, Antistin, Antistina, Antistine hydrochloride, Azalone, Ben-a-hist, Dibistin, Fenazolina, Histazine, Histostab, Imidamine, Phenazoline hydrochloride
IUPAC Name N-benzyl-N-(4,5-dihydro-1H-imidazol-2-ylmethyl)aniline;hydrochloride
SMILES Cl.C(N(Cc1ccccc1)c2ccccc2)C3=NCCN3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C17H19N3.ClH/c1-3-7-15(8-4-1)13-20(14-17-18-11-12-19-17)16-9-5-2-6-10-16;/h1-10H,11-14H2,(H,18,19);1H
Formula Weight 301.1346
Chemical Name or Material Antazoline hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.