missing translation for 'onlineSavingsMsg'
Learn More

Anthracene-9-boronic acid, 97%, Thermo Scientific Chemicals

Product Code. 10260070 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10260070

Brand: Thermo Scientific Acros 432812500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100622-34-2
Molecular Formula C14H11BO2
Molecular Weight (g/mol) 222.05
MDL Number MFCD03425925
InChI Key VHHDLIWHHXBLBK-UHFFFAOYSA-N
Synonym 9-anthraceneboronic acid, anthracene-9-boronic acid, 9-anthracenylboronic acid, 9-anthrylboronic acid, 9-anthracene boronic acid, boronic acid, 9-anthracenyl, 9-boronoanthracene, 9-borono-anthracene, zlchem 414, pubchem7757
PubChem CID 15510213
IUPAC Name anthracen-9-ylboronic acid
SMILES OB(O)C1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

Melting Point 215°C to 218°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Formula Weight 222.05
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material Anthracene-9-boronic acid
compliance-icons
Product Identifier
  • Anthracene-9-boronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.