missing translation for 'onlineSavingsMsg'
Learn More

Anthracene-9-carboxylic acid, 99%

Product Code. 10449010 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 10449010

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 723-62-6 | C15H10O2 | 222.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 723-62-6
Melting Point 213.0°C to 219.0°C
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C15H10O2
MDL Number MFCD00001257
Quantity 10 g
Beilstein 09, 705
Synonym 9-anthracenecarboxylic acid, 9-anthroic acid, anca, 9-carboxyanthracene, 9-ac, anthracene-10-carboxylic acid, 9-anthracene carboxylic acid, a9c, spectrum_001457, tocris-0963
Solubility Information Solubility in water: insoluble.
InChI Key XGWFJBFNAQHLEF-UHFFFAOYSA-N
SMILES OC(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12
IUPAC Name anthracene-9-carboxylic acid
Molecular Weight (g/mol) 222.24
PubChem CID 2201
ChEBI CHEBI:34507
Formula Weight 222.24
Percent Purity 99%
Chemical Name or Material Anthracene-9-carboxylic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Anthracene-9-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.