Learn More
Anthracene, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Anthracene is used as a source of dyestuffs and involved in the preparation of red dye alizarin. It is an organic semiconductor used as a scintillator for detectors of high energy photons, electrons and alfa particles. It is doped with polyvinyltoluene to prepare plastic scintillator. Further, it is used in the preparation of anthraquinone, wood preservatives, coating materials and pigments. In addition to this, it is used to prepare 1-hydroxyanthracene, 2-hydroxyanthracene and in 9,10-dihydroxyanthracene.
Solubility
Soluble in methanol, benzene, chloroform, ether, carbon disulfide, carbon tetrachloride and toluene. Slightly soluble in acetone. Insoluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents and hypochlorites.
Spécification
Spécification
| Melting Point | 213°C to 216°C |
| Boiling Point | 340°C to 342°C |
| Flash Point | 121°C (249°F) |
| Quantity | 250 g |
| UN Number | UN3077 |
| Beilstein | 1905429 |
| Merck Index | 14,682 |
| Formula Weight | 178.23 |
| Percent Purity | 97% |
| Chemical Name or Material | Anthracene |
Sécurité et traitement
- Anthracene
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H410-Very toxic to aquatic life with long lasting effects.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P391-Collect spillage.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Anthracene
RUO – Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.