missing translation for 'onlineSavingsMsg'
Learn More

Anthranilamide, 99+%

Product Code. 10050650 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10050650 100 g Plastic bottle 100g
10569610 5 g Glass bottle 5g
10530214 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10050650 Supplier Thermo Scientific Acros Supplier No. 104901000
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 88-68-6 | C7H8N2O | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-68-6
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00007981
InChI Key PXBFMLJZNCDSMP-UHFFFAOYSA-N
Synonym anthranilamide, benzamide, 2-amino, o-aminobenzamide, 2-carbamoylaniline, aminobenzamide, anthranilimidic acid, benzamide, o-amino, anthranilic acid amide, anthranilamide van, o-aminobenzamide van
PubChem CID 6942
IUPAC Name 2-aminobenzamide
SMILES C1=CC=C(C(=C1)C(=O)N)N

Specifications

Melting Point 111.0°C to 114.0°C
Boiling Point 300.0°C
Flash Point 198°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Plastic bottle
Linear Formula 2-(H2N)C6H4CONH2
Quantity 100 g
Beilstein 14, 320
Solubility Information Solubility in water: <5g/L water (20°C). Other solubilities: 140g/L ethanol (20°C), slightly soluble in ethylether, benzene
Formula Weight 136.15
Percent Purity 99+%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Anthranilamide
Show More Show Less
compliance-icons
Product Identifier
  • Anthranilamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.