missing translation for 'onlineSavingsMsg'
Learn More

Anthranilic Acid, Specified Reagent for general laboratory work, Fisher Chemical

Product Code. 11453523 Shop All Fisher Chemical Products
Change view
Click to view available options
:
100g, Glass Jar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11453523 100g, Glass Jar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 11453523 Supplier Fisher Chemical Supplier No. A/7680/48
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Specifications

CAS 118-92-3
Melting Point 144°C
Color Yellow
pH 4
Packaging Glass Jar
Molecular Formula C7H7NO2
MDL Number 7712
Quantity 100 g
Formula Weight 137.14g/mol
Physical Form Powder
compliance-icons
Product Identifier
  • ANTHRANILIC ACID
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.