missing translation for 'onlineSavingsMsg'
Learn More

Anthraquinone-2-carboxylic acid, 98%

Product Code. p-7041000
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15457428

Brand: Thermo Scientific Alfa Aesar H60929.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 117-78-2 | C15H8O4 | 252.225 g/mol

Anthraquinone 2-carboxylic acid (AQ) as a novel electron shuttling mediator and attached electron relay for HRP. Efficient method for the generation of hydrogen peroxide by aerobic photooxidation of 2-propanol using anthraquinone-2-carboxylic acid and molecular oxygen is investigated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anthraquinone 2-carboxylic acid (AQ) as a novel electron shuttling mediator and attached electron relay for HRP. Efficient method for the generation of hydrogen peroxide by aerobic photooxidation of 2-propanol using anthraquinone-2-carboxylic acid and molecular oxygen is investigated.

Solubility
Soluble in hot Acetic acid(almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-78-2
Molecular Formula C15H8O4
Molecular Weight (g/mol) 252.225
MDL Number MFCD00001231
InChI Key ASDLSKCKYGVMAI-UHFFFAOYSA-N
Synonym anthraquinone-2-carboxylic acid, 9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid, 2-carboxyanthraquinone, 2-anthraquinonecarboxylic acid, 2-carboxyl-9,10-anthraquinone, anthraquinone-2-carboxylate, 9,10-dihydro-9,10-dioxo-2-anthracenecarboxylic acid, 9,10-anthraquinone 2-carboxylic acid, .beta.-anthraquinonecarboxylic acid, anthraquinone-.beta.-carboxylic acid
PubChem CID 67030
IUPAC Name 9,10-dioxoanthracene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)C(=O)O

Specifications

Melting Point 287°C to 289°C
Quantity 1 g
Solubility Information Soluble in hot Acetic acid(almost transparency).
Formula Weight 252.22
Percent Purity 98%
Chemical Name or Material Anthraquinone-2-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.