missing translation for 'onlineSavingsMsg'
Learn More

Anthraquinone, TRC

Product Code. 30136801
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136801 250 g 250g
30134951 500 g 500g
17939339 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136801 Supplier Toronto Research Chemicals Supplier No. TRCA679245250G
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-65-1
Molecular Formula C14 H8 O2
Molecular Weight (g/mol) 208.21
Synonym Anthraquinone (8CI), 1,4,11,12-Tetrahydro-9,10-anthraquinone, 9,10-Anthraquinone, Anthracene-9,10-quinone, Anthradione, Avipel, DAQ-N, Flight Control, Flight Control Plus, Hoelite, Kawasaki SAQ, Morkit, NSC 7957, SAQ, SAQ (quinone)
IUPAC Name anthracene-9,10-dione
SMILES O=C1c2ccccc2C(=O)c3ccccc13

Specifications

Quantity 250 g
InChI Formula InChI=1S/C14H8O2/c15-13-9-5-1-2-6-10(9)14(16)12-8-4-3-7-11(12)13/h1-8H
Formula Weight 208.0524
Chemical Name or Material Anthraquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.