missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 61 publication
274.00 € - 555.00 €
Specifications
| Antigen | Caspase-8 |
|---|---|
| Dilution | Western Blot 1:1000-1:2000, Simple Western reported by internal validation, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18700284
|
Novus Biologicals
NB100-56116SS |
0.025 mL |
274.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18796363
|
Novus Biologicals
NB100-56116 |
0.05 mL |
555.00 €
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caspase-8 Polyclonal specifically detects Caspase-8 in Human, Mouse, Rat, Gerbil samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Caspase-8 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Apoptosis Detections, Cancer, Caspases, Cell Biology, Phospho Specific, Ubiquitin Proteasome Pathway | |
| Whole antisera with 0.05% Sodium Azide | |
| AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy | |
| CASP8 | |
| IgG | |
| Unpurified | |
| Detects Caspase-8 and cleavage products. |
| Western Blot 1:1000-1:2000, Simple Western reported by internal validation, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen 1:10-1:500 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat, Gerbil | |
| Q14790 | |
| 841 | |
| Recombinant catalytically active human Caspase-8 protein. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| 55.4 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title