Learn More
anti-AKT1, RPS6KB1 Protein Protein Interaction Antibody Pair, Abnova™
Description
Includes
AKT1 rabbit purified polyclonal antibody (20 ug); RPS6KB1 mouse monoclonal antibody (40 ug); Reagents are sufficient for at least 30-50 assays using recommended protocols
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the AKT1 protein, and the other against the RPS6KB1 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
Specifications
| Antigen | AKT1 |
| Applications | In situ PLA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Gene | AKT1 |
| Gene Alias | AKT/MGC99656/PKB/PKB-ALPHA/PRKBA/RAC/RAC-ALPHA |
| Gene Symbols | AKT1 |
| Host Species | Mouse, Rabbit |
| Quantity | 1 Set |
| Regulatory Status | RUO |
| Show More |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.