missing translation for 'onlineSavingsMsg'
Learn More
Learn More
arachidonate 12-lipoxygenase, 12R type, Mouse, Polyclonal Antibody, Abnova™
Description
This gene encodes an enzyme involved in the converstion of arachidonic acid to 12R-hydroxyeicosatetraenoic acid. Mutations in this gene are associated with nonbullous congenital ichthyosiform erythroderma. [provided by RefSeq
Sequence: NPNRPEWNGYIPGFPILINFKATKFLNLNLRYSFLKTASFFVRLGPMALAFKVRGLLDCKHSWKRLKDIRKIFPGKKSVVSEYVAEHWAED
Specifications
Specifications
| Antigen | arachidonate 12-lipoxygenase, 12R type |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant ALOX12B. |
| Formulation | 50% glycerol |
| Gene | ALOX12B |
| Gene Accession No. | NM_001139 |
| Gene Alias | 12R-LOX |
| Gene Symbols | ALOX12B |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?