missing translation for 'onlineSavingsMsg'
Learn More

Human Angiopoietin-like Protein 2/ANGPTL2 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

209.00 € - 2030.00 €

Specifications

Antigen Angiopoietin-like Protein 2/ANGPTL2
Clone 239829
Concentration LYOPH
Dilution Western Blot 1 ug/mL
Applications Western Blot
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18715769
GREENER_CHOICE
View Documents
R&D Systems
MAB2084-SP
25 μg
209.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18714388
GREENER_CHOICE
View Documents
R&D Systems
MAB2084-100
100 μg
513.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18759478
GREENER_CHOICE
View Documents
R&D Systems
MAB2084-500
500 μg
2030.00 €
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Angiopoietin-like Protein 2/ANGPTL2 Monoclonal specifically detects Angiopoietin-like Protein 2/ANGPTL2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Angiopoietin-like Protein 2/ANGPTL2
LYOPH
Western Blot
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
angiopoietin-like 2, Angiopoietin-like protein 2, angiopoietin-related protein 2, ANGPTL2, ANGRP2, ARP2HARP, MGC8889
ANGPTL2
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
239829
Western Blot 1 ug/mL
Monoclonal
Mouse
Human
Q9UKU9
23452
Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-like 2 Ile233-Phe492 Accession # Q9UKU9
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Angiopoietin-like 2 in direct ELISAs and Western blots. Shows 25-100% cross-reactivity with recombinant mouse Angiopoietin-like 2 and no cross-reactivity with recombinant human (rh) Angiopoietin-1 or rhAngiopoietin-2.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.