missing translation for 'onlineSavingsMsg'
Learn More

AP180 Mouse, Unlabeled, Clone: 34, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610469

Product Code. 15875479

  • ONLINE EXCLUSIVE PRICE
    265.00 €
    253.48 € / 1 set
    Save 11.52 €
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AP180, also known as AP-3, F1-20, NP185, and pp155, is one of four assembly proteins which are involved in the organization and assembly of clathrin triskelia in clathrin-coated vesicles. Two of these proteins, AP-1 and AP-2, exist as tetramers, while AP180 and auxilin are monomeric assembly proteins. The clathrin binding potential of AP180 is defined by several regions of the molecule, including the 30 kDa N-terminal domain, a central domain, and a 58 kDa C-terminal domain. Although clathrin binding occurs throughout the molecule, only the C-terminal domain is associated with both binding of clathrin and assembly of clathrin cages. The highly acidic central domain, which contains an uncharged alanine-rich segment, is thought to impart the irregular physical properties to this protein. AP180 is the only clathrin assembly protein specific for synapses and is thought to be involved in synaptic vesicle biogenesis and recycling. Furthermore, AP180 is bound by inositol-6-phosphate which has been shown to be closely regulated in neuronal cells by external stimuli.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

AP180
Monoclonal
250μg/mL
AP-3, F1-20, NP185, and pp155
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
34
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50 μg
Cell Biology
Human, Mouse, Rat
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.