missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATM Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 92 publications
228.00 € - 546.00 €
Specifications
| Antigen | ATM |
|---|---|
| Concentration | 2.5 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Flow Cytometry, ELISA, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunoblotting reported in scientific literature (PMID 28512243) |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18770354
|
Novus Biologicals
NB100-104SS |
0.025 mL |
228.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18797343
|
Novus Biologicals
NB100-104 |
546.00 €
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
ATM Polyclonal specifically detects ATM in Human, Mouse, Rat, Canine, Kangaroo samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting.Specifications
| ATM | |
| Western Blot 1:500-1:1000, Flow Cytometry, ELISA, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunoblotting reported in scientific literature (PMID 28512243) | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Genes Sensitive to DNA Damaging Agents, Hypoxia, p53 Pathway, Tumor Suppressors | |
| PBS, 50% Glycerol with 0.05% Sodium Azide | |
| AT mutated, A-T mutated, AT1, ATA, ataxia telangiectasia mutated (includes complementation groups A, C and D), ataxia telangiectasia mutatedATD, ATC, ATDC, ATE, DKFZp781A0353, EC 2.7.11.1, MGC74674, serine-protein kinase ATM, TEL1, TEL1, telomere maintenance 1, homolog, TELO1 | |
| ATM | |
| IgG | |
| Affinity Purified | |
| 350 kDa |
| 2.5 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Canine, Kangaroo | |
| Q13315 | |
| 472 | |
| A fragment of the human ATM protein corresponding to the C-terminus (within the last third of the protein sequence). [Uniprot: Q13315] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title