missing translation for 'onlineSavingsMsg'
Learn More

Bcl-2 w/control Hamster anti-Human, Unlabeled, Clone: 6C8, BD

Hamster Monoclonal Antibody

Brand:  BD Biosciences 551051

 View more versions of this product

Product Code. 15856488

  • ONLINE EXCLUSIVE PRICE
    285.00 €
    273.70 € / Each
    Save 11.30 €
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Bcl-2 is considered to be novel among proto-oncogenes because it blocks apoptosis (programmed cell death) in many cell types. Apoptosis is an active form of cellular suicide that typically requires new RNA and protein synthesis and is associated with distinct morphological changes including cell shrinkage, cytoplasm membrane blebbing, nuclear fragmentation and DNA degradation. The Bcl-2 gene was first found in t(14:18) containing follicular B-cell lymphomas. A high proportion of these lymphomas contains t(14:18) chromosomal translocations involving the human Bcl-2 gene. Translocation of Bcl-2 sequences from chromosome 18 onto the transcriptionally active immunoglobulin locus at chromosome band 14q32 in B-cells deregulates Bcl-2 gene expression, resulting in high levels of Bcl-2 mRNA and protein expression. Because Bcl-2 blocks apoptosis, it may contribute to tumorigenesis by prolonged cell survival rather than by accelerating the rate of cell proliferation. The reduced molecular weight of Bcl-2 is 26 kDa. Additional minor bands at 27-31 kDa and 18-21 kDa may also be observed.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bcl-2 w/control
Monoclonal
Unconjugated
Affinity Purified
RUO
Human
IgG
Western Blot
6C8
Hamster
50 μg
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.