missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCL7C Polyclonal Antibody

Description
Recommended positive controls: Jurkat. Predicted reactivity: Mouse (91%), Rat (92%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is identified by the similarity of its product to the N-terminal region of BCL7A protein. The BCL7A protein is encoded by the gene known to be directly involved in a three-way gene translocation in a Burkitt lymphoma cell line. The function of this gene has not yet been determined.
Specifications
Specifications
| Antigen | BCL7C |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | BCL7C |
| Gene Accession No. | Q8WUZ0 |
| Gene Alias | B cell CLL/lymphoma 7C; B-cell chronic lymphocytic leukemia/lymphoma 7C protein; B-cell CLL/lymphoma 7 protein family member C; B-cell CLL/lymphoma 7C; BCL tumor suppressor 7C; Bcl7c; C230096E12Rik |
| Gene Symbols | BCL7C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?