missing translation for 'onlineSavingsMsg'
Learn More

Bub3 Mouse, Unlabeled, Clone: 31, BD

Product Code. 10386744
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10386744 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10386744 Supplier BD Biosciences Supplier No. 611730

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Accurate chromosome segregation requires that all pairs of sister chromatids become appropriately attached to mitotic spindles before the onset of anaphase. Cell cycle checkpoints monitor kinetochore-microtubule interactions, so that cell cycle progression can be delayed until proper chromosome attachments are formed. In yeast, Bub1-3 genes are required for proper mitotic delay in response to unattached kinetochores. In mammals, the homologues to yeast Bub1 and Bub3 form a complex that binds kinetochores and has protein kinase activity. Bub3 contains four WD repeats, three in the N-terminus and one in the C-terminus, and a central Bub1-binding domain. During prophase and prometaphase, Bub3 localizes to the kinetochore before attachment to microtubules. In addition, taxol-induced formation of lagging chromosomes due to a delay of cell cycle progression increases the level of Bub3 co-localized with kinetochores, while correctly aligned chromosomes found in metaphase do not exhibit this co-localization. Thus, Bub3, in association with Bub1, may be important for sensing kinetochore attachment to microtubules during the prometaphase to metaphase transition.

Host Species: Mouse
Clone: 31
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human Bub3 aa. 4-16

Bioimaging, Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Bub3
Applications Western Blot
Classification Monoclonal
Clone 31
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Bub3 aa. 4-16
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.