missing translation for 'onlineSavingsMsg'
Learn More

c-Cbl Mouse, Unlabeled, Clone: 17, BD

Product Code. 15885459
Change view
Click to view available options
Quantity:
150 μg
50 μg
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15885459 50 μg 1 set
15895459 150 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15885459 Supplier BD Biosciences Supplier No. 610441

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

cbl was identified in the genome of a transforming retrovirus from a mouse pre-B cell lymphoma. The product of mammalian c-cbl, p120[cbl], is a widely expressed cytoplasmic protein with several distinctive domains including a RING finger motif and a large proline-rich domain. Although the normal cellular function of c-Cbl has not yet been determined, it associates with many well known signal transduction proteins. In lymphoid cells, c-Cbl is a substrate of TCR-activated protein tyrosine kinases. In addition, stimulation with EGF, PDGF, FGF, or NGF results in tyrosine-phosphorylation of c-Cbl in lymphoid and other cell types. It forms a signaling complex with Grb2 in T cells and is a SH3 and SH2 binding protein which associates with many other cytoplasmic signaling proteins including Btk, Src, fyn, lck, and the β isoform of PI-3 kinase. Thus, c-Cbl may be an important adaptor protein in growth factor and T-cell receptor signaling pathways.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Cbl
Applications Western Blot
Classification Monoclonal
Clone 17
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.