Learn More
anti-CAPS Matched Antibody Pair, Abnova
Brand: Abnova H00000828-AP21.1Set
Includes:
- 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-CAPS (100μg)
- 2. Detection antibody: mouse purified polyclonal anti-CAPS (20μg)
- Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This gene encodes a calcium-binding protein, which may play a role in the regulation of ion transport. A similar protein was first described as a potentially important regulatory protein in the dog thyroid and was termed as R2D5 antigen in rabbit. Alternative splicing of this gene generates two transcript variants.
- Species Reactivity: Human
ELISA
Specifications
| CAPS | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CAPS. | |
| CAPS1/MGC126562 | |
| Mouse, Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-CAPS (100 μg) 2. Detection antibody: mouse purified polyclonal anti-CAPS (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| CAPS | |
| CAPS | |
| NA; CAPS (NP_004049.1, 1 a.a. ∽ 189 a.a) full-length human protein. | |
| 1 Set | |
| Signal Transduction | |
| 828 | |
| Human |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.