missing translation for 'onlineSavingsMsg'
Learn More

Carboxypeptidase E Mouse, Unlabeled, Clone: 35, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610759

 View more versions of this product

Product Code. 15835629

  • ONLINE EXCLUSIVE PRICE
    476.00 €
    456.78 € / 1 set
    Save 19.22 €
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Carboxypeptidase E (CPE), also known as carboxypeptidase H and enkephalin convertase, is found as both a membrane-bound and a soluble glycoprotein in neuroendocrine tissues and adrenal-gland chromaffin granules. The C-terminus forms an amphiphilic α-helix, suggesting that this region is responsible for the membrane-bound form. Evidence suggests the active form of CPE is located in the secretory vesicles. CPE appears to have several functions. It is an exopeptidase that cleaves neuropeptides with C-terminal basic amino acids, producing an active form of the peptide. It has also been proposed that membrane-bound CPE is a sorting receptor for regulated secretory pathway (RSP) proteins in the TGN pituitary Golgi and secretory granule membranes. RSP proteins primarily consist of hormones and neuropeptides. Mice that carry a mutation in the CPE gene Cpe[fat] display endocrine disorders such as obesity, infertility, and hyperproinsulinemia. Furthermore, the same endocrine disorders are observed in Cpe[fat] mice where the CPE gene has been effaced by antisense RNA.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Carboxypeptidase E
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human Carboxypeptidase E aa. 49-200
150 μg
Cell Biology
Human, Mouse, Rat
IgG1
Western Blot
35
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.